COM448686247 Combi-Blocks, Inc.

1-benzyl 4-methyl (2S)-2-{[(benzyloxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578298
  • Cas Number : 1820579-34-7
  • Molecular Weight : 371.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0771

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0771
Availability details for
Quantity
  • Availability
  • Add to Cart