COM448686251 Combi-Blocks, Inc.

methyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoate

  • MDL number MFCD27578301
  • Cas Number : 1820579-45-0
  • Molecular Weight : 367.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0781
Availability details for
Quantity
  • Availability
  • Add to Cart