COM448686254 Combi-Blocks, Inc.

(S)-2-tert-Butoxycarbonylamino-3-naphthalen-2-yl-propionic acid methyl ester

  • MDL number MFCD11501455
  • Cas Number : 176896-73-4
  • Molecular Weight : 329.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0790
Availability details for
Quantity
  • Availability
  • Add to Cart