COM448686256 Combi-Blocks, Inc.

Dimethyl 3-thienylmalonate

  • MDL number MFCD08436111
  • Cas Number : 70158-52-0
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0795

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0795
Availability details for
Quantity
  • Availability
  • Add to Cart