COM448686259 Combi-Blocks, Inc.

Methyl sinapate

  • MDL number MFCD00210467
  • Cas Number : 20733-94-2
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0798
Availability details for
Quantity
  • Availability
  • Add to Cart