COM448686264 Combi-Blocks, Inc.

methyl 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylpropanoate

  • MDL number MFCD27578303
  • Cas Number : 203636-40-2
  • Molecular Weight : 339.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0806
Availability details for
Quantity
  • Availability
  • Add to Cart