COM448686271 Combi-Blocks, Inc.

1,4-dimethyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578305
  • Cas Number : 147220-14-2
  • Molecular Weight : 383.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0822
Availability details for
Quantity
  • Availability
  • Add to Cart