COM448686272 Combi-Blocks, Inc.

4-tert-butyl 1-methyl (2R)-2-{[(benzyloxy)carbonyl]amino}butanedioate hydrate

  • MDL number MFCD27578306
  • Cas Number : 1820580-02-6
  • Molecular Weight : 355.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0826
Availability details for
Quantity
  • Availability
  • Add to Cart