COM448686273 Combi-Blocks, Inc.

Methyl 3-(chloromethyl)benzoate

  • MDL number MFCD00157184
  • Cas Number : 34040-63-6
  • Molecular Weight : 184.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0829
Availability details for
Quantity
  • Availability
  • Add to Cart