COM448686275 Combi-Blocks, Inc.

(3-Chloro-4-fluoro-phenyl)-acetic acid methyl ester

  • MDL number MFCD12546087
  • Cas Number : 1189770-43-1
  • Molecular Weight : 202.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0836

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0836
Availability details for
Quantity
  • Availability
  • Add to Cart