COM448686282 Combi-Blocks, Inc.

(1R,2R)-cyclohexane-1,2-diamine 1,4-dimethyl (2R,3R)-2,3-dihydroxybutanedioate

  • MDL number MFCD27578308
  • Cas Number : 1820575-00-5
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0862
Availability details for
Quantity
  • Availability
  • Add to Cart