COM448686293 Combi-Blocks, Inc.

Methyl 2-(trifluoromethyl)cinnamate

  • MDL number MFCD04973007
  • Cas Number : 157518-53-1
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0887

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0887
Availability details for
Quantity
  • Availability
  • Add to Cart