COM448686294 Combi-Blocks, Inc.

1-(9H-Fluoren-9-ylmethyl) 3-methyl piperidine-1,3-dicarboxylate

  • MDL number MFCD24227432
  • Cas Number : 1552171-24-0
  • Molecular Weight : 365.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0888
Availability details for
Quantity
  • Availability
  • Add to Cart