COM448686295 Combi-Blocks, Inc.

Boc-D-Thr-OMe

  • MDL number MFCD01073763
  • Cas Number : 96099-84-2
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0889
Availability details for
Quantity
  • Availability
  • Add to Cart