COM448686297 Combi-Blocks, Inc.

methyl 2,3,4,9-tetrahydro-1H-carbazole-3-carboxylate

  • MDL number MFCD27578315
  • Cas Number : 26088-67-5
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0891

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0891
Availability details for
Quantity
  • Availability
  • Add to Cart