COM448686300 Combi-Blocks, Inc.

methyl 2-fluoro-3-(trifluoromethoxy)benzoate

  • MDL number MFCD27578318
  • Cas Number : 1803825-64-0
  • Molecular Weight : 238.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0894
Availability details for
Quantity
  • Availability
  • Add to Cart