COM448686304 Combi-Blocks, Inc.

(S)-2-Amino-3-(4-hydroxy-3-nitro-phenyl)-propionic acid methyl ester

  • MDL number MFCD09031837
  • Cas Number : 3195-65-1
  • Molecular Weight : 240.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0901
Availability details for
Quantity
  • Availability
  • Add to Cart