COM448686306 Combi-Blocks, Inc.

Methyl 3-(4-chlorophenyl)propanoate

  • MDL number MFCD08273468
  • Cas Number : 50561-69-8
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0914

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0914
Availability details for
Quantity
  • Availability
  • Add to Cart