COM448686308 Combi-Blocks, Inc.

1-(2,4-Difluoro-phenyl)-5-oxo-pyrrolidine-3-carboxylic acid methyl ester

  • MDL number MFCD16630905
  • Cas Number : 1239766-16-5
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0917
Availability details for
Quantity
  • Availability
  • Add to Cart