COM448686314 Combi-Blocks, Inc.

methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}pentanoate

  • MDL number MFCD27578320
  • Cas Number : 116611-58-6
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0927
Availability details for
Quantity
  • Availability
  • Add to Cart