COM448686317 Combi-Blocks, Inc.

3-(4-Bromo-2-fluoro-phenyl)-propionic acid methyl ester

  • MDL number MFCD18398780
  • Cas Number : 1257107-61-1
  • Molecular Weight : 261.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0939

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0939
Availability details for
Quantity
  • Availability
  • Add to Cart