COM448686325 Combi-Blocks, Inc.

Carbobenzyloxyglycylglycine methyl ester

  • MDL number MFCD00158470
  • Cas Number : 13437-63-3
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0956
Availability details for
Quantity
  • Availability
  • Add to Cart