COM448686328 Combi-Blocks, Inc.

methyl (2S)-6-{[(tert-butoxy)carbonyl]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoate

  • MDL number MFCD27578324
  • Cas Number : 172846-56-9
  • Molecular Weight : 482.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0960

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0960
Availability details for
Quantity
  • Availability
  • Add to Cart