COM448686329 Combi-Blocks, Inc.

methyl (2R)-2-{[(tert-butoxy)carbonyl](methyl)amino}-3-methylbutanoate

  • MDL number MFCD27578325
  • Cas Number : 1268729-59-4
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0963

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0963
Availability details for
Quantity
  • Availability
  • Add to Cart