COM448686330 Combi-Blocks, Inc.

methyl 5-fluoro-2-[4-fluoro-2-(methoxycarbonyl)phenyl]benzoate

  • MDL number MFCD27578326
  • Cas Number : 1820640-40-1
  • Molecular Weight : 306.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0965
Availability details for
Quantity
  • Availability
  • Add to Cart