COM448686332 Combi-Blocks, Inc.

Z-Lys-OMe

  • MDL number MFCD10001332
  • Cas Number : 5591-93-5
  • Molecular Weight : 294.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0972
Availability details for
Quantity
  • Availability
  • Add to Cart