COM448686333 Combi-Blocks, Inc.

(3-Bromo-4-fluorophenyl)acetic acid methyl ester

  • MDL number MFCD12025009
  • Cas Number : 885681-61-8
  • Molecular Weight : 247.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0973
Availability details for
Quantity
  • Availability
  • Add to Cart