COM448686334 Combi-Blocks, Inc.

(S)-Tetrahydrofuran-2-carboxylic acid methyl ester

  • MDL number MFCD00211270
  • Cas Number : 87324-01-4
  • Molecular Weight : 130.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0975

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0975
Availability details for
Quantity
  • Availability
  • Add to Cart