COM448686337 Combi-Blocks, Inc.

methyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[(triphenylmethyl)carbamoyl]propanoate

  • MDL number MFCD27578329
  • Cas Number : 1500080-56-7
  • Molecular Weight : 610.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0981

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0981
Availability details for
Quantity
  • Availability
  • Add to Cart