COM448686344 Combi-Blocks, Inc.

1-benzyl 4-methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578330
  • Cas Number : 158201-15-1
  • Molecular Weight : 337.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1007
Availability details for
Quantity
  • Availability
  • Add to Cart