COM448686346 Combi-Blocks, Inc.

4-tert-butyl 1-methyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578331
  • Cas Number : 172846-53-6
  • Molecular Weight : 425.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1015

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1015
Availability details for
Quantity
  • Availability
  • Add to Cart