COM448686347 Combi-Blocks, Inc.

methyl 3-methoxy-4-(2-methylpropoxy)benzoate

  • MDL number MFCD09741645
  • Cas Number : 3535-29-3
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1017
Availability details for
Quantity
  • Availability
  • Add to Cart