COM448686348 Combi-Blocks, Inc.

1-tert-butyl 5-methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}pentanedioate

  • MDL number MFCD27578332
  • Cas Number : 24277-38-1
  • Molecular Weight : 317.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1018

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1018
Availability details for
Quantity
  • Availability
  • Add to Cart