COM448686349 Combi-Blocks, Inc.

methyl (2S)-1-(2-{[(tert-butoxy)carbonyl]amino}acetyl)pyrrolidine-2-carboxylate

  • MDL number MFCD27578333
  • Cas Number : 41863-49-4
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1019
Availability details for
Quantity
  • Availability
  • Add to Cart