COM448686350 Combi-Blocks, Inc.

Benzoic acid, 3-borono-5-(1,1-dimethylethyl)-, 1-methyl ester

  • MDL number MFCD23380359
  • Cas Number : 193905-91-8
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1020

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1020
Availability details for
Quantity
  • Availability
  • Add to Cart