COM448686353 Combi-Blocks, Inc.

methyl 2-(isopropylsulfanyl)benzoate

  • MDL number MFCD27578334
  • Cas Number : 154550-03-5
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1027
Availability details for
Quantity
  • Availability
  • Add to Cart