COM448686354 Combi-Blocks, Inc.

4-tert-butyl 1-methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578335
  • Cas Number : 18635-49-9
  • Molecular Weight : 303.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1030
Availability details for
Quantity
  • Availability
  • Add to Cart