COM448686358 Combi-Blocks, Inc.

Biphenyl 3-carboxylic acid methyl ester

  • MDL number MFCD01464091
  • Cas Number : 16606-00-1
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1042

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1042
Availability details for
Quantity
  • Availability
  • Add to Cart