COM448686362 Combi-Blocks, Inc.

methyl 2-(5-chloro-3-fluoropyridin-2-yl)acetate

  • MDL number MFCD27578338
  • Cas Number : 1804879-61-5
  • Molecular Weight : 203.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1055
Availability details for
Quantity
  • Availability
  • Add to Cart