COM448686367 Combi-Blocks, Inc.

3,4-dimethyl 2-hydroxy-6-methylpyridine-3,4-dicarboxylate

  • MDL number MFCD27578342
  • Cas Number : 41516-33-0
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1063

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1063
Availability details for
Quantity
  • Availability
  • Add to Cart