COM448686368 Combi-Blocks, Inc.

Methyl (2r)-2-acetamido-4-methylpentanoate

  • MDL number MFCD24549479
  • Cas Number : 35799-87-2
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1064

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1064
Availability details for
Quantity
  • Availability
  • Add to Cart