COM448686370 Combi-Blocks, Inc.

1-(2,4-Dichloro-phenyl)-5-oxo-pyrrolidine-3-carboxylic acid methyl ester

  • MDL number MFCD16630877
  • Cas Number : 1239769-46-0
  • Molecular Weight : 288.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1070

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1070
Availability details for
Quantity
  • Availability
  • Add to Cart