COM448686371 Combi-Blocks, Inc.

Methyl 4-ethoxy-3-methoxybenzoate

  • MDL number MFCD06203685
  • Cas Number : 3535-24-8
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1071

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1071
Availability details for
Quantity
  • Availability
  • Add to Cart