COM448686373 Combi-Blocks, Inc.

Methyl (2s)-2-amino-3-(4-iodophenyl)propanoate

  • MDL number MFCD08058398
  • Cas Number : 113850-77-4
  • Molecular Weight : 305.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1077
Availability details for
Quantity
  • Availability
  • Add to Cart