COM448686374 Combi-Blocks, Inc.

methyl (2S)-3-[4-(tert-butoxy)phenyl]-2-acetamidopropanoate

  • MDL number MFCD27578345
  • Cas Number : 1820580-42-4
  • Molecular Weight : 293.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1078
Availability details for
Quantity
  • Availability
  • Add to Cart