COM448686378 Combi-Blocks, Inc.

Methyl (s)-(-)-4-amino-2-hydroxybutanoate

  • MDL number MFCD19206592
  • Cas Number : 140156-44-1
  • Molecular Weight : 133.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1091

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1091
Availability details for
Quantity
  • Availability
  • Add to Cart