COM448686380 Combi-Blocks, Inc.

(R)-Methyl 2-methoxy-2-phenylacetate

  • MDL number MFCD16879060
  • Cas Number : 32174-46-2
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1094
Availability details for
Quantity
  • Availability
  • Add to Cart