COM448686382 Combi-Blocks, Inc.

methyl 1-{[(benzyloxy)carbonyl]amino}cyclopentane-1-carboxylate

  • MDL number MFCD27578348
  • Cas Number : 1820674-23-4
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1096
Availability details for
Quantity
  • Availability
  • Add to Cart