COM448686383 Combi-Blocks, Inc.

Methyl 2-([(1s)-1-phenylethyl]amino)acetate

  • MDL number MFCD18903630
  • Cas Number : 248603-82-9
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1097
Availability details for
Quantity
  • Availability
  • Add to Cart