COM448686384 Combi-Blocks, Inc.

Methyl 3-carbamimidamidopropanoate

  • MDL number MFCD16739982
  • Cas Number : 1249472-28-3
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1098
Availability details for
Quantity
  • Availability
  • Add to Cart